Assay Details
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256081.2 | XP_006256143.1 | 1 - 2 | 157 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
