Assay Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256065.2 | XP_006256127.1 | 1 - 2 | 80 | 82 | |
GenBank mRNA | D82069.1 | - | 1 - 2 | 85 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D82069.1, XM_006256065.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
