Assay Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256062.2 | XP_006256124.1 | 2 - 3 | 191 | 71 | |
XM_006256063.2 | XP_006256125.1 | 2 - 3 | 154 | 71 | ||
XM_006256065.2 | XP_006256127.1 | 3 - 4 | 269 | 71 | ||
GenBank mRNA | D82069.1 | - | 3 - 4 | 274 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D82069.1, XM_006256062.2, XM_006256063.2, XM_006256065.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
