Assay Details
Gene Symbol: Stk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013197.2 | NP_001013215.2 | 2 - 3 | 284 | 102 | |
GenBank mRNA | CO398403.1 | - | 2 - 3 | 277 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
