Assay Details
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212458.1 | NP_997623.1 | 3 - 4 | 585 | 64 | |
XM_006255972.1 | XP_006256034.1 | 3 - 4 | 438 | 64 | ||
XM_006255973.1 | XP_006256035.1 | 2 - 3 | 339 | 64 | ||
XM_006255974.1 | XP_006256036.1 | 3 - 4 | 527 | 64 | ||
XM_006255975.1 | XP_006256037.1 | 3 - 4 | 828 | 64 | ||
GenBank mRNA | BC085852.1 | - | 3 - 4 | 531 | 7189891 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
