Assay Details
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212458.1 | NP_997623.1 | 4 - 5 | 767 | 102 | |
XM_006255972.1 | XP_006256034.1 | 4 - 5 | 620 | 102 | ||
XM_006255973.1 | XP_006256035.1 | 3 - 4 | 521 | 102 | ||
XM_006255974.1 | XP_006256036.1 | 4 - 5 | 709 | 102 | ||
XM_006255975.1 | XP_006256037.1 | 4 - 5 | 1010 | 102 | ||
GenBank mRNA | BC085852.1 | - | 4 - 5 | 713 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
