Assay Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109025.2 | NP_001102495.1 | 2 - 3 | 261 | 74 | |
GenBank mRNA | BC088341.1 | - | 2 - 3 | 237 | 7367801 | 74 |
DY573537.1 | - | 2 - 3 | 261 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
