Assay Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109025.2 | NP_001102495.1 | 3 - 4 | 367 | 144 | |
GenBank mRNA | BC088341.1 | - | 3 - 4 | 343 | 7367801 | 144 |
DY573537.1 | - | 3 - 4 | 367 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
