Assay Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109025.2 | NP_001102495.1 | 5 - 6 | 545 | 92 | |
GenBank mRNA | BC088341.1 | - | 5 - 6 | 521 | 92 | |
DY573537.1 | - | 5 - 6 | 545 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
