Assay Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109137.1 | NP_001102607.1 | 13 - 14 | 4522 | 70 | |
XM_006227887.2 | XP_006227949.1 | 14 - 15 | 6875 | 70 | ||
XM_008758736.1 | XP_008756958.1 | 14 - 15 | 6359 | 70 | ||
XM_008758737.1 | XP_008756959.1 | 14 - 15 | 7047 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P144423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25106 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144423 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144423 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P144423 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P258343 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99246 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39440 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10702871 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702872 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702875 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702866 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702874 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702868 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702879 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10702865 | - | - |
Rat Genome 230 2.0 Array | 1382068_at | - | - |
Rat Genome 230 2.0 Array | 1382068_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382068_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382068_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100002991.rn.1 | TC0100000487.rn.1 | RTA TC |
Rat Expression Set 230 | 1382068_at | - | - |
Rat Expression Set 230 | 1382068_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702865 | - | - |
Rat Genome U34 Set | rc_AA963991_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21087144 | - | - |
ABI Rat Genome Survey Microarray | 21305871 | - | - |
ABI Rat Genome Survey Microarray | 21808738 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360830 | - | - |
