Assay Details
Gene Symbol: Tmem181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130939.1 | NP_001124411.1 | 1 - 2 | 110 | 86 | |
GenBank mRNA | BC168231.1 | - | 1 - 2 | 110 | 86 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168231.1, NM_001130939.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
