Assay Details
Gene Symbol: Tmem181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130939.1 | NP_001124411.1 | 5 - 6 | 478 | 100 | |
GenBank mRNA | BC168231.1 | - | 5 - 6 | 478 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
