Assay Details
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127560.1 | NP_001121032.1 | 1 - 2 | 296 | 91 | |
XM_006227888.2 | XP_006227950.1 | 2 - 3 | 849 | 91 | ||
XM_006227891.2 | XP_006227953.1 | 2 - 3 | 849 | 91 | ||
GenBank mRNA | BC166706.1 | - | 1 - 2 | 296 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
