Assay Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109137.1 | NP_001102607.1 | 5 - 6 | 861 | 78 | |
XM_006227887.2 | XP_006227949.1 | 6 - 7 | 3214 | 78 | ||
XM_008758736.1 | XP_008756958.1 | 6 - 7 | 2698 | 78 | ||
XM_008758737.1 | XP_008756959.1 | 6 - 7 | 3386 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
