Assay Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109137.1 | NP_001102607.1 | 9 - 10 | 1634 | 89 | |
XM_006227887.2 | XP_006227949.1 | 10 - 11 | 3987 | 89 | ||
XM_008758736.1 | XP_008756958.1 | 10 - 11 | 3471 | 89 | ||
XM_008758737.1 | XP_008756959.1 | 10 - 11 | 4159 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
