Assay Details
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019357.1 | NP_062230.1 | 12 - 13 | 2077 | 69 | |
GenBank mRNA | BC081958.1 | - | 12 - 13 | 2077 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
leukocyte cell-cell adhesion
regulation of cell shape
positive regulation of gene expression
membrane to membrane docking
microvillus assembly
epithelial cell differentiation
receptor internalization
establishment or maintenance of apical/basal cell polarity
establishment of epithelial cell apical/basal polarity
filopodium assembly
actin filament bundle assembly
establishment of endothelial barrier
RNA binding
structural molecule activity
protein binding
protein domain specific binding
protein-containing complex binding
cell adhesion molecule binding
actin filament binding
leukocyte cell-cell adhesion
regulation of cell shape
positive regulation of gene expression
membrane to membrane docking
microvillus assembly
epithelial cell differentiation
receptor internalization
establishment or maintenance of apical/basal cell polarity
establishment of epithelial cell apical/basal polarity
filopodium assembly
actin filament bundle assembly
establishment of endothelial barrier
RNA binding
structural molecule activity
protein binding
protein domain specific binding
protein-containing complex binding
cell adhesion molecule binding
actin filament binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P809953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70940 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P809953 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P809953 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P809953 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15737 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P809953 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700540825 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28826 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63588 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84565 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700540825 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10717894 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717900 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717902 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717901 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717896 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717899 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717892 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717904 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717897 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717895 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717898 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717906 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717893 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717903 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10717891 | - | - |
Rat Genome 230 2.0 Array | 1370875_at | - | - |
Rat Genome 230 2.0 Array | 1370875_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370875_at | - | - |
Affymetrix HT Rat Focus Array | 1370875_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100035228.rn.1 | TC0100004426.rn.1 | RTA TC |
Rat Expression Set 230 | 1370875_PM_at | - | - |
Rat Expression Set 230 | 1370875_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717891 | - | - |
Rat Genome U34 Set | X67788_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21775859 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361603 | - | - |
RatRef-12 Expression BeadChip | ILMN_69560 | - | - |
