Assay Details
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019357.1 | NP_062230.1 | 3 - 4 | 680 | 140 | |
GenBank mRNA | AF450298.1 | - | 3 - 4 | 279 | 140 | |
AY428869.1 | - | 3 - 4 | 265 | 140 | ||
BC081958.1 | - | 3 - 4 | 680 | 7124292 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
