Assay Details
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113544.1 | NP_001107016.1 | 1 - 2 | 116 | 104 | |
XM_006227892.2 | XP_006227954.1 | 1 - 2 | 162 | 104 | ||
GenBank mRNA | CB613741.1 | - | 1 - 2 | 111 | 104 | |
CB613959.1 | - | 1 - 2 | 116 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001113544.1, XM_006227892.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
