Assay Details
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113542.1 | NP_001107014.1 | 4 - 5 | 339 | 139 | |
NM_001113543.1 | NP_001107015.1 | 4 - 5 | 339 | 139 | ||
NM_001113544.1 | NP_001107016.1 | 4 - 5 | 273 | 139 | ||
XM_006227892.2 | XP_006227954.1 | 4 - 5 | 319 | 139 | ||
XM_006227893.2 | XP_006227955.1 | 4 - 5 | 497 | 139 | ||
GenBank mRNA | CB613741.1 | - | 4 - 5 | 268 | 139 | |
CB613959.1 | - | 4 - 5 | 273 | 139 | ||
CK478322.1 | - | 4 - 5 | 339 | 139 | ||
CV108433.1 | - | 4 - 5 | 336 | 139 | ||
FQ228682.1 | - | 4 - 5 | 378 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
