Assay Details
Gene Symbol: Tmem181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130939.1 | NP_001124411.1 | 15 - 16 | 1383 | 86 | |
GenBank mRNA | BC168231.1 | - | 15 - 16 | 1383 | 86 | |
FQ211485.1 | - | 935 | 86 | |||
FQ228553.1 | - | 288 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
