Assay Details
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012468.1 | NP_001012486.1 | 3 - 4 | 233 | 149 | |
XM_006254132.2 | XP_006254194.1 | 3 - 4 | 290 | 149 | ||
GenBank mRNA | BC089957.1 | - | 3 - 4 | 233 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
