Assay Details
Gene Symbol: Cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012941.2 | NP_037073.1 | 1 - 2 | 237 | 112 | |
GenBank mRNA | AY724494.1 | - | 1 - 2 | 559 | 112 | |
BC087033.1 | - | 1 - 2 | 237 | 112 | ||
D55681.1 | - | 1 - 2 | 290 | 112 | ||
U17697.1 | - | 1 - 2 | 403 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
