Assay Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014151.2 | NP_001014173.2 | 7 - 8 | 989 | 60 | |
XM_006254965.2 | XP_006255027.1 | 5 - 6 | 700 | 60 | ||
XM_006254966.2 | XP_006255028.1 | 5 - 6 | 1153 | 60 | ||
GenBank mRNA | AY539873.1 | - | 7 - 8 | 989 | 60 | |
BC078941.1 | - | 5 - 6 | 674 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
