Assay Details
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023093.1 | NP_075581.1 | 1 - 2 | 109 | 134 | |
GenBank mRNA | BC061830.1 | - | 1 - 2 | 109 | 5599635 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061830.1, NM_023093.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
