Assay Details
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108728.1 | NP_001102198.1 | 10 - 11 | 1836 | 142 | |
XM_006240766.2 | XP_006240828.1 | 11 - 12 | 1956 | 142 | ||
GenBank mRNA | FQ220010.1 | - | 7 - 8 | 931 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
