Assay Details
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108728.1 | NP_001102198.1 | 5 - 6 | 1173 | 85 | |
XM_006240760.2 | XP_006240822.1 | 6 - 7 | 1078 | 85 | ||
XM_006240761.2 | XP_006240823.1 | 6 - 7 | 1090 | 85 | ||
XM_006240762.2 | XP_006240824.1 | 7 - 8 | 1198 | 85 | ||
XM_006240763.2 | XP_006240825.1 | 7 - 8 | 1186 | 85 | ||
XM_006240764.2 | XP_006240826.1 | 5 - 6 | 982 | 85 | ||
XM_006240766.2 | XP_006240828.1 | 6 - 7 | 1293 | 85 | ||
XM_006240767.2 | XP_006240829.1 | 4 - 5 | 742 | 85 | ||
XM_008765027.1 | XP_008763249.1 | 6 - 7 | 1112 | 85 | ||
XM_008765028.1 | XP_008763250.1 | 6 - 7 | 1078 | 85 | ||
GenBank mRNA | FQ220010.1 | - | 2 - 3 | 268 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
