Assay Details
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108728.1 | NP_001102198.1 | 6 - 7 | 1316 | 98 | |
XM_006240760.2 | XP_006240822.1 | 7 - 8 | 1221 | 98 | ||
XM_006240761.2 | XP_006240823.1 | 7 - 8 | 1233 | 98 | ||
XM_006240762.2 | XP_006240824.1 | 8 - 9 | 1341 | 98 | ||
XM_006240763.2 | XP_006240825.1 | 8 - 9 | 1329 | 98 | ||
XM_006240764.2 | XP_006240826.1 | 6 - 7 | 1125 | 98 | ||
XM_006240766.2 | XP_006240828.1 | 7 - 8 | 1436 | 98 | ||
XM_006240767.2 | XP_006240829.1 | 5 - 6 | 885 | 98 | ||
XM_008765027.1 | XP_008763249.1 | 7 - 8 | 1255 | 98 | ||
XM_008765028.1 | XP_008763250.1 | 7 - 8 | 1221 | 98 | ||
GenBank mRNA | FQ220010.1 | - | 3 - 4 | 411 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
