Assay Details
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077677.1 | NP_001071145.1 | 1 - 2 | 376 | 116 | |
GenBank mRNA | BC127534.1 | - | 1 - 2 | 376 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
