Assay Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 11 - 12 | 954 | 62 | |
XM_006244296.2 | XP_006244358.1 | 11 - 12 | 1012 | 62 | ||
XM_008766751.1 | XP_008764973.1 | 12 - 13 | 1470 | 62 | ||
GenBank mRNA | AF482705.1 | - | 11 - 12 | 954 | 62 | |
BC086988.1 | - | 11 - 12 | 965 | 7192831 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
