Assay Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 6 - 7 | 500 | 85 | |
XM_006244296.2 | XP_006244358.1 | 6 - 7 | 558 | 85 | ||
XM_008766746.1 | XP_008764968.1 | 7 - 8 | 1016 | 85 | ||
XM_008766747.1 | XP_008764969.1 | 668 | 85 | |||
XM_008766748.1 | XP_008764970.1 | 7 - 8 | 1001 | 85 | ||
XM_008766749.1 | XP_008764971.1 | 6 - 7 | 560 | 85 | ||
XM_008766750.1 | XP_008764972.1 | 6 - 7 | 545 | 85 | ||
XM_008766751.1 | XP_008764973.1 | 7 - 8 | 1016 | 85 | ||
XM_008766752.1 | XP_008764974.1 | 6 - 7 | 908 | 85 | ||
XM_008766753.1 | XP_008764975.1 | 6 - 7 | 893 | 85 | ||
GenBank mRNA | AF482705.1 | - | 6 - 7 | 500 | 85 | |
BC086988.1 | - | 6 - 7 | 511 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
