Assay Details
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_217313.8 | XP_217313.6 | 1 - 2 | 1903 | 76 | |
GenBank mRNA | FQ231323.1 | - | 1 - 2 | 190 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
