Assay Details
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343671.6 | XP_343672.2 | 12 - 13 | 2336 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
