Assay Details
Gene Symbol: Shd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108223.1 | NP_001101693.1 | 1 - 2 | 211 | 70 | |
XM_006244329.1 | XP_006244391.1 | 1 - 2 | 2297 | 70 | ||
GenBank mRNA | BC158646.1 | - | 1 - 2 | 153 | 8364383 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC158646.1, NM_001108223.1, XM_006244329.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
