Assay Details
Gene Symbol: Shd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108223.1 | NP_001101693.1 | 2 - 3 | 553 | 85 | |
GenBank mRNA | BC158646.1 | - | 2 - 3 | 495 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
