Assay Details
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031239.2 | NP_112518.1 | 9 - 10 | 1050 | 54 | |
GenBank mRNA | AB008161.1 | - | 9 - 10 | 1050 | 54 | |
AF009602.1 | - | 9 - 10 | 1035 | 54 | ||
BC070893.1 | - | 9 - 10 | 1046 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
