Assay Details
Gene Symbol: Hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054007.1 | NP_446459.1 | 4 - 5 | 394 | 169 | |
GenBank mRNA | AF035156.1 | - | 4 - 5 | 394 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
