Assay Details
Gene Symbol: Hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054007.1 | NP_446459.1 | 10 - 11 | 833 | 73 | |
GenBank mRNA | AF035156.1 | - | 10 - 11 | 833 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
organic acid metabolic process
steroid biosynthetic process
response to herbicide
response to metal ion
response to lead ion
response to organic cyclic compound
response to activity
response to insecticide
insecticide metabolic process
biphenyl metabolic process
phenol-containing compound metabolic process
hippocampus development
response to nutrient levels
cellular response to hormone stimulus
response to vitamin E
Leydig cell differentiation
response to L-ascorbic acid
response to estrogen
response to ethanol
response to arsenic-containing substance
response to cadmium ion
response to steroid hormone
response to corticosterone
response to fungicide
testosterone biosynthetic process
response to fatty acid
cellular response to antibiotic
cellular response to gonadotropin stimulus
estradiol 17-beta-dehydrogenase activity
oxidoreductase activity
testosterone dehydrogenase (NAD+) activity
testosterone 17-beta-dehydrogenase (NADP+) activity
organic acid metabolic process
steroid biosynthetic process
response to herbicide
response to metal ion
response to lead ion
response to organic cyclic compound
response to activity
response to insecticide
insecticide metabolic process
biphenyl metabolic process
phenol-containing compound metabolic process
hippocampus development
response to nutrient levels
cellular response to hormone stimulus
response to vitamin E
Leydig cell differentiation
response to L-ascorbic acid
response to estrogen
response to ethanol
response to arsenic-containing substance
response to cadmium ion
response to steroid hormone
response to corticosterone
response to fungicide
testosterone biosynthetic process
response to fatty acid
cellular response to antibiotic
cellular response to gonadotropin stimulus
estradiol 17-beta-dehydrogenase activity
oxidoreductase activity
testosterone dehydrogenase (NAD+) activity
testosterone 17-beta-dehydrogenase (NADP+) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62150 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146105 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146105 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146105 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13021 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17374 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94894 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66364 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10793525 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793522 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793520 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793517 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793524 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793518 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793519 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793521 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10793516 | - | - |
Rat Genome 230 2.0 Array | 1369553_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369553_at | - | - |
Affymetrix HT Rat Focus Array | 1369553_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369553_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700008112.rn.1 | TC1700001181.rn.1 | RTA TC |
Rat Expression Set 230 | 1369553_PM_at | - | - |
Rat Expression Set 230 | 1369553_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793516 | - | - |
Rat Genome U34 Set | AF035156_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20863149 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369450 | - | - |
RatRef-12 Expression BeadChip | ILMN_66918 | - | - |
