Assay Details
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004203.3 | NP_001004203.3 | 3 - 4 | 466 | 83 | |
XM_006249005.2 | XP_006249067.1 | 3 - 4 | 588 | 83 | ||
XM_006249006.2 | XP_006249068.1 | 3 - 4 | 471 | 83 | ||
GenBank mRNA | BC078973.1 | - | 3 - 4 | 466 | 7111847 | 83 |
DQ278199.1 | - | 3 - 4 | 514 | 83 | ||
DQ278200.1 | - | 1059 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
