Assay Details
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044225.1 | NP_001037690.1 | 8 - 9 | 894 | 153 | |
XM_008769061.1 | XP_008767283.1 | 8 - 9 | 934 | 153 | ||
XM_008769062.1 | XP_008767284.1 | 8 - 9 | 933 | 153 | ||
GenBank mRNA | BC100108.1 | - | 8 - 9 | 894 | 153 | |
D49446.1 | - | 7 - 8 | 865 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
