Assay Details
Gene Symbol: Slc35a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127642.1 | NP_001121114.1 | 2 - 3 | 289 | 118 | |
XM_006256693.2 | XP_006256755.1 | 2 - 3 | 571 | 118 | ||
GenBank mRNA | BC166811.1 | - | 2 - 3 | 289 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
