Assay Details
Gene Symbol: Slc35a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127642.1 | NP_001121114.1 | 3 - 4 | 443 | 112 | |
XM_006256693.2 | XP_006256755.1 | 3 - 4 | 725 | 112 | ||
GenBank mRNA | BC166811.1 | - | 3 - 4 | 443 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
