Assay Details
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008298.1 | NP_001008299.1 | 4 - 5 | 480 | 156 | |
GenBank mRNA | BC086352.1 | - | 4 - 5 | 480 | 156 | |
FQ220233.1 | - | 4 - 5 | 503 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
