Assay Details
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199502.1 | NP_955796.1 | 10 - 11 | 1592 | 111 | |
XM_006257373.2 | XP_006257435.1 | 10 - 11 | 1619 | 111 | ||
GenBank mRNA | AB080636.1 | - | 10 - 11 | 1592 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
