Assay Details
Gene Symbol: Tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109014.1 | NP_001102484.1 | 2 - 3 | 727 | 75 | |
XM_008773426.1 | XP_008771648.1 | 2 - 3 | 756 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
