Assay Details
Gene Symbol: Tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109014.1 | NP_001102484.1 | 5 - 6 | 924 | 71 | |
XM_008773426.1 | XP_008771648.1 | 5 - 6 | 953 | 71 | ||
XM_008773427.1 | XP_008771649.1 | 4 - 5 | 812 | 71 | ||
XM_008773428.1 | XP_008771650.1 | 4 - 5 | 350 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
