Assay Details
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133538.2 | NP_598222.2 | 2 - 3 | 217 | 71 | |
GenBank mRNA | AF448818.1 | - | 2 - 3 | 217 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF448818.1, NM_133538.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
