Assay Details
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012762.2 | NP_036894.2 | 3 - 4 | 450 | 97 | |
GenBank mRNA | D85899.1 | - | 3 - 4 | 450 | 97 | |
FQ228818.1 | - | 1 - 2 | 82 | 97 | ||
U14647.1 | - | 3 - 4 | 450 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
