Assay Details
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106798.2 | NP_001100268.2 | 5 - 6 | 787 | 71 | |
GenBank mRNA | BC098729.1 | - | 4 - 5 | 1143 | 71 | |
BC166800.1 | - | 5 - 6 | 787 | 71 | ||
FQ209549.1 | - | 5 - 6 | 803 | 71 | ||
FQ214028.1 | - | 5 - 6 | 806 | 71 | ||
FQ214274.1 | - | 5 - 6 | 800 | 71 | ||
FQ214451.1 | - | 5 - 6 | 801 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
