Assay Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108989.1 | NP_001102459.1 | 1 - 2 | 48 | 74 | |
XM_008765202.1 | XP_008763424.1 | 1 - 2 | 75 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108989.1, XM_008765202.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
