Assay Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108989.1 | NP_001102459.1 | 2 - 3 | 106 | 131 | |
XM_008765202.1 | XP_008763424.1 | 2 - 3 | 133 | 131 | ||
GenBank mRNA | BC166847.1 | - | 2 - 3 | 89 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
